Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP2 Antibody (MT-01), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB50415A750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB50415A750 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB50415A750 Supplier Novus Biologicals Supplier No. NB500415AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MAP2 Monoclonal specifically detects MAP2 in Human, Mouse, Porcine, Bovine, Feline samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen MAP2
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone MT-01
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias DKFZp686I2148, MAP-2, MAP2A, MAP2B, MAP2C, Microtubule Associated Protein 2, microtubule-associated protein 2, MTAP2
Gene Symbols MAP2
Host Species Mouse
Immunogen MAP2 Antibody (MT-01) was developed against the Thermostabile fraction of bovine brain microtubules (whole MAP2ab molecule).
Quantity 0.1 mL
Research Discipline Cell Biology, Cellular Markers, MAP Kinase Signaling, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4133
Test Specificity The antibody MT-01 reacts with molecule Microtubule Associated Protein 2ab (MAP2ab).
Target Species Human, Mouse, Pig, Bovine, Feline
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.