Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP2 Antibody, Novus Biologicals™
SDP

Catalog No. NB017943 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017943 0.025 mL
NB10098717 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017943 Supplier Novus Biologicals Supplier No. NB100987170.025ML
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

MAP2 Polyclonal specifically detects MAP2 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen MAP2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:300-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:500, Immunohistochemistry-Paraffin 1:300-1:2000
Formulation Whole antisera with No Preservative
Gene Accession No. P20357
Gene Alias DKFZp686I2148, MAP-2, MAP2A, MAP2B, MAP2C, microtubule-associated protein 2
Gene Symbols MAP2
Host Species Sheep
Immunogen A synthetic peptide as a part of rat, mouse Mtap2 conjugated to an immunogenic carrier protein was used as the immunogen.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Cytoskeleton Markers, MAP Kinase Signaling, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4133
Test Specificity MAP-2.
Reconstitution Reconstitute with 0.1 ml sterilized water to desired concentration.
Target Species Rat, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.