Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP3K12 Antibody, Novus Biologicals™
SDP

Catalog No. NB018602 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018602 0.025 mg
NBP14606305 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018602 Supplier Novus Biologicals Supplier No. NBP1460630.025MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 6 publications

MAP3K12 Polyclonal specifically detects MAP3K12 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunoprecipitation.

Specifications

Antigen MAP3K12
Applications Immunoprecipitation, Western Blot, ELISA
Classification Polyclonal
Concentration 0.5mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:1000, Immunoprecipitation 1:200
Formulation Tris/Glycine buffer pH 7.5-7.9, stabilizing protein, and glycerol with 0.02% Sodium Azide
Gene Accession No. Q12852
Gene Alias DLKEC 2.7.11.25, Dual leucine zipper bearing kinase, EC 2.7.11, Leucine-zipper protein kinase, MAPK-upstream kinase, MEKK12, mitogen-activated protein kinase kinase kinase 12, Mixed lineage kinase, MUKdual leucine zipper kinase DLK, protein kinase MUK, ZPKleucine zipper protein kinase, ZPKP1
Gene Symbols MAP3K12
Host Species Rabbit
Immunogen Synthetic peptides corresponding to unique epitope on MAP3K12. The peptide sequence was selected from mid-region, amimo acids 270-320. This peptide was covalently modified post-synthetically to achieve desired antigenicity.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 7786
Test Specificity Specifically detects a unique epitope on MAP3K12.
Target Species Rat, Mouse, Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.