Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP3K15 Antibody (OTI1G10), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP272588G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272588G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272588G Supplier Novus Biologicals Supplier No. NBP272588G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MAP3K15 Monoclonal specifically detects MAP3K15 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen MAP3K15
Applications Western Blot
Classification Monoclonal
Clone OTI1G10
Conjugate DyLight 488
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Apoptosis signal-regulating kinase 3, ASK3MEKK 15, bA723P2.3, EC 2.7.11, EC 2.7.11.25, FLJ16518, MAPK/ERK kinase kinase 15, MEK kinase 15, mitogen-activated protein kinase kinase kinase 15
Gene Symbols MAP3K15
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 979-1313 of human MAP3K15 (NP_001001671) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 389840.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.