Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP3K15 Antibody (OTI1G10), PerCP, Novus Biologicals™
SDP

Catalog No. NB287994 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287994 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB287994 Supplier Novus Biologicals Supplier No. NBP272588PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MAP3K15 Monoclonal antibody specifically detects MAP3K15 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen MAP3K15
Applications Western Blot
Classification Monoclonal
Clone OTI1G10
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias Apoptosis signal-regulating kinase 3, ASK3MEKK 15, bA723P2.3, EC 2.7.11, EC 2.7.11.25, FLJ16518, MAPK/ERK kinase kinase 15, MEK kinase 15, mitogen-activated protein kinase kinase kinase 15
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 979-1313 of human MAP3K15(NP_001001671) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 389840
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.