Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP3K8/Tpl2/COT Antibody (OTI3D9), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. N272589A647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272589A647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272589A647 Supplier Novus Biologicals Supplier No. NBP272589AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MAP3K8/Tpl2/COT Monoclonal specifically detects MAP3K8/Tpl2/COT in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen MAP3K8/Tpl2/COT
Applications Western Blot
Classification Monoclonal
Clone OTI3D9
Conjugate Alexa Fluor 647
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Cancer Osaka thyroid oncogene, c-COT, cot (cancer Osaka thyroid) oncogene, COTMEKK8, EC 2.7.11.25, ESTF, ESTFLJ10486, Ewing sarcoma transformant, mitogen-activated protein kinase kinase kinase 8, Proto-oncogene c-Cot, proto-oncogene serine/threoine protein kinase, Serine/threonine-protein kinase cot, Tpl-2, TPL2, Tumor progression locus 2, tumor progression locus-2
Gene Symbols MAP3K8
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 277-461 of human MAP3K8/Tpl2/COT (NP_005195) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Angiogenesis, Cancer, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 1326.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.