Learn More
Description
Specifications
Specifications
| Antigen | MAP4K2 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | B lymphocyte serine/threonine protein kinase, B lymphocyte serine/threonine-protein kinase, BL44, EC 2.7.11, EC 2.7.11.1, GC kinase, GCKMEK kinase kinase 2, Germinal center kinase, germinal centre kinase (GC kinase), MAPK/ERK kinase kinase kinase 2, mitogen-activated protein kinase kinase kinase kinase 2, Rab8 interacting protein, Rab8-interacting protein, RAB8IPMEKKK 2 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 551-820 of human MAP4K2 (NP_004570.2). |
| Purification Method | Affinity purified |
| Quantity | 100 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
