Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP4K2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB093511 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB093511 20 μL
NB093510 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB093511 Supplier Novus Biologicals Supplier No. NBP30510520UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MAP4K2 Polyclonal specifically detects MAP4K2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MAP4K2
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias B lymphocyte serine/threonine protein kinase, B lymphocyte serine/threonine-protein kinase, BL44, EC 2.7.11, EC 2.7.11.1, GC kinase, GCKMEK kinase kinase 2, Germinal center kinase, germinal centre kinase (GC kinase), MAPK/ERK kinase kinase kinase 2, mitogen-activated protein kinase kinase kinase kinase 2, Rab8 interacting protein, Rab8-interacting protein, RAB8IPMEKKK 2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 551-820 of human MAP4K2 (NP_004570.2).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5871
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.