Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP4K3 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB327073 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB327073 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB327073 Supplier Novus Biologicals Supplier No. NBP299100F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MAP4K3 Polyclonal antibody specifically detects MAP4K3 in Human samples. It is validated for Western Blot

Specifications

Antigen MAP4K3
Applications Western Blot
Classification Polyclonal
Conjugate FITC
Dilution Western Blot
Formulation PBS
Gene Alias EC 2.7.11, EC 2.7.11.1, germinal center kinase-like kinase, Germinal center kinase-related protein kinase, GLKMEKKK 3, MAPK/ERK kinase kinase kinase 3, MAPKKKK3, MEK kinase kinase 3, mitogen-activated protein kinase kinase kinase kinase 3, RAB8IPL1
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human MAP4K3.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 8491
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.