Learn More
Description
Specifications
Specifications
| Antigen | MAP4K3 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | EC 2.7.11, EC 2.7.11.1, germinal center kinase-like kinase, Germinal center kinase-related protein kinase, GLKMEKKK 3, MAPK/ERK kinase kinase kinase 3, MAPKKKK3, MEK kinase kinase 3, mitogen-activated protein kinase kinase kinase kinase 3, RAB8IPL1 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 755-894 of human MAP4K3 (NP_003609.2). |
| Purification Method | Affinity purified |
| Quantity | 100 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
