Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP4K6 Antibody, Novus Biologicals™
SDP

Catalog No. NBP122990 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP122990 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP122990 Supplier Novus Biologicals Supplier No. NBP122990
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

MAP4K6 Polyclonal specifically detects MAP4K6 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen MAP4K6
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. Q8N4C8
Gene Alias B55, EC 2.7.11, GCK family kinase MiNK, hMINK, hMINKbeta, MAP4K6EC 2.7.11.1, MAPK/ERK kinase kinase kinase 6, MEK kinase kinase 6, MGC21111, MINKMEKKK 6, Misshapen/NIK-related kinase, misshapen-like kinase 1, misshapen-like kinase 1 (zebrafish), Mitogen-activated protein kinase kinase kinase kinase 6, serine/threonine protein kinase, YSK2, ZC3
Gene Symbols MINK1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 775 and 825 of human misshapen-like kinase 1 using the numbering given in entry NP_722549.2 (GeneID 50488).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, MAP Kinase Signaling, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 50488
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, Crab-eating macaque, Chinese hamster, Small-eared galago, Thirteen-lined ground squirrel, Little brown bat and Northern white-cheeked gibbon.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.