Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPK15 Antibody, Novus Biologicals™
SDP

Catalog No. NBP156789 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP156789 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP156789 Supplier Novus Biologicals Supplier No. NBP156789
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MAPK15 Polyclonal specifically detects MAPK15 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen MAPK15
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunocytochemistry/Immunofluorescence 1:200
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q8TD08
Gene Alias EC 2.7.11, ERK-7, ERK7MAPK 15, ERK-8, ERK8EC 2.7.11.24, extracellular regulated kinase 8 delta, extracellular signal regulated kinase 8, Extracellular signal-regulated kinase 7, Extracellular signal-regulated kinase 8, MAP kinase 15, mitogen-activated protein kinase 15
Gene Symbols MAPK15
Host Species Rabbit
Immunogen Synthetic peptides corresponding to MAPK15(mitogen-activated protein kinase 15) The peptide sequence was selected from the middle region of MAPK15. Peptide sequence GHDPAEHESPRAAKNVPRQNSAPLLQTALLGNGERPPGAKEAPPLTLSLV.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225689
Test Specificity Expected identity based on immunogen sequence: Human: 100%;.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.