Learn More
MAPK1IP1L Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | MAPK1IP1L |
---|---|
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1129
![]() |
Bethyl Laboratories, Inc
A305743AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine and Orangutan
Mitogen-activated protein kinase 1-interacting protein 1-like (MAPK1IP1L) belongs to the MISS family.Specifications
MAPK1IP1L | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
C14orf32, MISS, c14_5346, MAPK-interacting and spindle-stabilizing protein, MAPK-interacting and spindle-stabilizing protein-like, mitogen activated protein kinase 1 interacting protein 1-like, mitogen-activated protein kinase 1-interacting protein 1-like | |
Between 1 and 50 | |
Primary | |
4° C | |
MAPK1IP1L |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q8BH93, Q8NDC0 | |
218975, 93487 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.