Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPKAP Kinase 3 Rabbit anti-Human, Mouse, Clone: JU30-31, Novus Biologicals™
SDP

Catalog No. NBP275565 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP275565 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP275565 Supplier Novus Biologicals Supplier No. NBP275565

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Monoclonal Antibody

MAPKAP Kinase 3 Monoclonal specifically detects MAPKAP Kinase 3 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen MAPKAP Kinase 3
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone JU30-31
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunoprecipitation 1:10 - 1:50
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias 3PKEC 2.7.11.1, Chromosome 3p kinase, EC 2.7.11, MAP kinase-activated protein kinase 3, MAPK-activated protein kinase 3, MAPKAP kinase 3, MAPKAP33pK, MAPKAPK-3, mitogen-activated protein kinase-activated protein kinase 3
Gene Symbols MAPKAPK3
Host Species Rabbit
Immunogen Synthetic peptide within Human MAPKAP Kinase 3 aa 351-400/ 400. (SwissProt: P49137 Human; SwissProt: P49138 Mouse)
Molecular Weight of Antigen 46 kDa
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 7867.0
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.