Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPKAPK2 Antibody (7H4.2) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB019457 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019457 0.05 mg
NBP229767 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019457 Supplier Novus Biologicals Supplier No. NBP2297670.05MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MAPKAPK2 Monoclonal specifically detects MAPKAPK2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen MAPKAPK2
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 7H4.2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunoprecipitation 1:20
Formulation 0.2 mg antibody with PBS with 0.1% Sodium Azide
Gene Accession No. P49137
Gene Alias EC 2.7.11, EC 2.7.11.1, MAP kinase-activated protein kinase 2, MAPK-activated protein kinase 2, MAPKAP kinase 2, MAPKAPK-2, mitogen-activated protein kinase-activated protein kinase 2, MK2
Gene Symbols MAPKAPK2
Host Species Mouse
Immunogen KLH-conjugated synthetic peptide of the C-terminus of the long form of human MAPKAP-K2.
Molecular Weight of Antigen 45 kDa
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, MAP Kinase Signaling, Phospho Specific, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9261
Target Species Human, Rat, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.