Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Marapsin/Pancreasin Antibody (MM0463-3H15), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP211786FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211786FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP211786FR Supplier Novus Biologicals Supplier No. NBP211786FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Marapsin/Pancreasin Monoclonal specifically detects Marapsin/Pancreasin in Human samples. It is validated for Western Blot.

Specifications

Antigen Marapsin/Pancreasin
Applications Western Blot
Classification Monoclonal
Clone MM0463-3H15
Conjugate DyLight 680
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CAPH2, channel-activating protease 2, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, EC 3.4.21.43, marapsin, MPNMarapsin, Pancreasin, protease, serine 27, serine protease 27
Gene Symbols PRSS27
Host Species Mouse
Immunogen Human recombinant Marapsin/Pancreasin
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 83886
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.