Learn More
Description
Specifications
Specifications
| Antigen | MARCH10 |
| Applications | Western Blot, Immunohistochemistry (Paraffin), ELISA |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000, Immunohistochemistry-Paraffin, Peptide ELISA Detection limit 1:8000 |
| Formulation | Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide |
| Gene Accession No. | NP_001094345.1 |
| Gene Alias | EC 6.3.2.-, FLJ35757, MARCH-Xprobable E3 ubiquitin-protein ligase MARCH10, membrane-associated ring finger (C3HC4) 10, Membrane-associated RING finger protein 10, Membrane-associated RING-CH protein X, RING finger protein 190RNF190 |
| Gene Symbols | 43169 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
