Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MARCH5 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15958520 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15958520 20 μL
NBP159585 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15958520 Supplier Novus Biologicals Supplier No. NBP15958520UL

Rabbit Polyclonal Antibody

44260 Polyclonal specifically detects 44260 in Human samples. It is validated for Western Blot, Immunoblotting, Knockout Validated.

Specifications

Antigen MARCH5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS & 2% Sucrose. with No Preservative
Gene Accession No. Q9NX47
Gene Alias E3 ubiquitin-protein ligase MARCH5, EC 6.3.2, EC 6.3.2.-, FLJ20445, MARCH-VRNF153, membrane-associated ring finger (C3HC4) 5, Membrane-associated RING finger protein 5, Membrane-associated RING-CH protein V, Mitochondrial ubiquitin ligase, RING finger protein 153MITOL
Gene Symbols 43164
Host Species Rabbit
Immunogen Synthetic peptides corresponding to MARCH5(membrane-associated ring finger (C3HC4) 5) The peptide sequence was selected form the N terminal of 40242. Peptide sequence CRGSTKWVHQACLQRWVDEKQRGNSTARVACPQCNAEYLIVFPKLGPVVY.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 54708
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.