Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MARCH8 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB356954 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB356954 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB356954 Supplier Novus Biologicals Supplier No. NBP176959MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MARCH8 Polyclonal antibody specifically detects MARCH8 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen MARCH8
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Cellular modulator of immune recognition, EC 6.3.2, EC 6.3.2.-, MARCH-VIIIcellular modulator of immune recognition, membrane-associated ring finger (C3HC4) 8, Membrane-associated RING finger protein 8, Membrane-associated RING-CH protein VIII, MIR, RING finger protein 178
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide from near the carboxy terminus of human MARCH8. The immunogen is located within the last 50 amino acids of MARCH8. Amino Acid Squence: CPETSKKNIFEKSPLTE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 220972
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.