Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MARCKS Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB360308 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB360308 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB360308 Supplier Novus Biologicals Supplier No. NBP305539APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MARCKS Monoclonal antibody specifically detects MARCKS in Human, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen MARCKS
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Conjugate APC
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias 80K-L, 80K-L protein, FLJ14368, MACSmyristoylated alanine-rich C-kinase substrate, MRACKS, myristoylated alanine-rich protein kinase C substrate, myristoylated alanine-rich protein kinase C substrate (MARCKS, 80K-L), phosphomyristin, PKCSLFLJ90045, PRKCSL, Protein kinase C substrate, 80 kDa protein, light chain
Host Species Mouse
Immunogen Full length human MARCKS expressed in and purified from E. coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4082
Target Species Human, Monkey
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.