Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MARCKS Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 1 publication

Supplier:  Novus Biologicals NB11058875SS

 View more versions of this product

Catalog No. NB11058875T


Only null left
Add to Cart

Description

Description

MARCKS Polyclonal specifically detects MARCKS in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

MARCKS
Polyclonal
Western Blot 1:10000-1:20000, Immunohistochemistry 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:500-1:5000
80K-L, 80K-L protein, FLJ14368, MACSmyristoylated alanine-rich C-kinase substrate, MRACKS, myristoylated alanine-rich protein kinase C substrate, myristoylated alanine-rich protein kinase C substrate (MARCKS, 80K-L), phosphomyristin, PKCSLFLJ90045, PRKCSL, Protein kinase C substrate, 80 kDa protein, light chain
Rabbit
80 kDa
0.025 mL
Primary
Human
Antisera
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Unconjugated
50% PBS, 50% glycerol with 5mM Sodium Azide
MARCKS
Full length recombinant rat MARCKS expressed in and purified from E. coli. [UniProt# P30009]
Unpurified
RUO
4082
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only