Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MARCKS Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 1 publication

$206.00

Specifications

Antigen MARCKS
Dilution Western Blot 1:10000-1:20000, Immunohistochemistry 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:500-1:5000
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB11058875T
SDP
View Documents
Novus Biologicals
NB11058875SS
0.025 mL
Each for $206.00
Only null left
Add to Cart
 
NB11058875
SDP
View Documents
Novus Biologicals
NB11058875
0.1 mL N/A N/A N/A
Description

Description

MARCKS Polyclonal specifically detects MARCKS in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

MARCKS
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Unconjugated
Rabbit
Human
80K-L, 80K-L protein, FLJ14368, MACSmyristoylated alanine-rich C-kinase substrate, MRACKS, myristoylated alanine-rich protein kinase C substrate, myristoylated alanine-rich protein kinase C substrate (MARCKS, 80K-L), phosphomyristin, PKCSLFLJ90045, PRKCSL, Protein kinase C substrate, 80 kDa protein, light chain
MARCKS
IgG
Unpurified
80 kDa
Western Blot 1:10000-1:20000, Immunohistochemistry 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:500-1:5000
Polyclonal
Antisera
RUO
50% PBS, 50% glycerol with 5mM Sodium Azide
4082
Full length recombinant rat MARCKS expressed in and purified from E. coli. [UniProt# P30009]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.