Learn More
Invitrogen™ MARCKSL1 Polyclonal Antibody

Description
Predicted to react with bovine, mouse, rabbit and rat based on sequence homology.
Specifications
Specifications
| Antigen | MARCKSL1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | Marcksl1 |
| Gene Accession No. | P49006 |
| Gene Alias | AL022768; AW215397; AW536807; brain protein F52; D4Bc1; F52; MACMARCKS; Mac-MARCKS; Macrophage myristoylated alanine-rich C kinase substrate; Macs2; Macs3; MARCKS like 1; Marcksl1; MARCKS-like 1; MARCKS-like protein 1; MARCKS-related protein; MLP; MLP1; Mrp; myristoylated alanine-rich C kinase substrate |
| Gene Symbols | Marcksl1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.