Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MARCO Antibody (2359A), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB348592 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB348592 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB348592 Supplier Novus Biologicals Supplier No. FAB29561MFV500
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

MARCO Monoclonal specifically detects MARCO in Mouse samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen MARCO
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 2359A
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias macrophage receptor with collagenous structureSCARA2Scavenger receptor class A member 2, member 2
Host Species Rabbit
Immunogen Mouse myeloma cell line, NS0-derived mouse MARCO protein, Gln70-Ser518, Accession # Q60754
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 8685
Target Species Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.