Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MARCO Antibody (579511), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB363555 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB363555 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB363555 Supplier Novus Biologicals Supplier No. FAB2956M
Add to Cart
Add to Cart

Rat Monoclonal Antibody

MARCO Monoclonal specifically detects MARCO in Mouse samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen MARCO
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 579511
Conjugate DyLight 594
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias macrophage receptor with collagenous structureSCARA2Scavenger receptor class A member 2, member 2
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse MARCO, Gln70-Ser518, Accession # Q60754
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 8685
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.