Learn More
Description
Specifications
Specifications
| Antigen | MARCO |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay |
| Classification | Monoclonal |
| Clone | PLK-1 |
| Concentration | 0.1 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Functional |
| Formulation | 0.2 um filtered solution in PBS, 0.1% BSA |
| Gene Alias | macrophage receptor with collagenous structureSCARA2Scavenger receptor class A member 2, member 2 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
