Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MARCO Monoclonal Antibody (PLK-1), PE-Cyanine7, eBioscience™
GREENER_CHOICE

Catalog No. p-7175404
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-544-742 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 25-544-742 Supplier Invitrogen™ Supplier No. 25544742
Only null left
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Description: This PLK-1 monoclonal antibody recognizes human MARCO also known as SR-A6 or SCARA2. Applications Reported: This PLK-1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This PLK-1 antibody has been pre-diluted and tested by flow cytometric analysis of human monocyte-derived macrophages. This may be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333-34) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.

MARCO, also known as SR-A6 or SCARA2, is a macrophage surface receptor and a member of the class A scavenger receptor family. As a part of the innate antimicrobial immune system, MARCO can bind both Gram-negative and Gram-positive bacteria via an extracellular, C-terminal, scavenger receptor cysteine-rich (SRCR) domain. In addition to short cytoplasmic and transmembrane domains, there is an extracellular spacer domain and a long, extracellular collagenous domain. The protein may form a trimeric molecule by the association of the collagenous domains of three identical polypeptide chains. Due to its expression in tumor infiltrating macrophages, and its role in inhibiting immune responses within tumor environment, MARCO has been indicated as a potential target for cancer immunotherapy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MARCO
Applications Flow Cytometry
Classification Monoclonal
Clone PLK-1
Concentration 5 μL/Test
Conjugate PE-Cyanine7
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene Marco
Gene Accession No. Q9UEW3
Gene Alias AI323439; Ly112; macrophage receptor MARCO; Macrophage receptor with collagenous structure; MARCO; SCARA2; Scavenger receptor class A member 2; scavenger receptor class A, member 2; SR-A6
Gene Symbols Marco
Host Species Mouse
Immunogen Human alveolar macrophages.
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8685
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.