Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Marimastat, 98%, PROMO, Thermo Scientific Chemicals

Catalog No. J67288MAPM
Click to view available options
Quantity:
10 mg

Chemical Identifiers

CAS 154039-60-8
Molecular Formula C15H29N3O5
Synonym (2S,3R)-N4-[(1S)-2,2-Dimethyl-1-[(methylamino)carbonyl] propyl]-N1,2-dihydroxy-3-(2-methylpropyl)butanediamide; BB2516

Specifications

Quantity 10 mg
Content And Storage -30°C to -10°C
Melting Point 148°C
Form Solid
Assay 98%

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.