Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Marimastat, 98%

Catalog Number AAJ67288MCR
Click to view available options
Quantity:
1 mg
10 mg
154039-60-8
C15H29N3O5
(2S,3R)-N4-[(1S)-2,2-Dimethyl-1-[(methylamino)carbonyl] propyl]-N1,2-dihydroxy-3-(2-methylpropyl)butanediamide; BB2516
154039-60-8
C15H29N3O5
(2S,3R)-N4-[(1S)-2,2-Dimethyl-1-[(methylamino)carbonyl] propyl]-N1,2-dihydroxy-3-(2-methylpropyl)butanediamide; BB2516

A broad spectrum inhibitor of matrix metalloproteases (MMPs); inhibits MMP-9, MMP-1, MMP-2, MMP-14 and MMP-7 with IC50 values of 3, 5, 6, 9 and 13 nM respectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 mg
Description White
Content And Storage -30°C to -10°C
Formula Weight 331.41g/mol
Melting Point 148°C
Form Solid
InChI Key OCSMOTCMPXTDND-OUAUKWLOSA-N
SMILES CNC(=O)[C@@H](NC(=O)[C@H](CC(C)C)[C@H](O)C(=O)NO)C(C)(C)C
IUPAC Name (2S,3R)-N'-[(1S)-2,2-dimethyl-1-(methylcarbamoyl)propyl]-N,2-dihydroxy-3-(2-methylpropyl)butanediamide
Molecular Weight (g/mol) 331.41
Assay 98%
Show More Show Less
Recommended Storage Store at -20°C

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit