Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MARK2 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB291039 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291039 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291039 Supplier Novus Biologicals Supplier No. NBP171889APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MARK2 Polyclonal antibody specifically detects MARK2 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen MARK2
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias EC 2.7.11, EC 2.7.11.1, ELKL motif kinase, ELKL motif kinase 1, EMK-1, EMK1PAR-1, MAP/microtubule affinity-regulating kinase 2MGC99619, PAR1 homolog, Par1b, Ser/Thr protein kinase PAR-1B, serine/threonine protein kinase EMK, serine/threonine-protein kinase MARK2
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 300 and 350 of human MAP/microtubule Affinity-Regulating Kinase 2 using the numbering given in entry NP_001034558.2 (GeneID 2011).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2011
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.