| Antigen |
MARK2 (p Thr595) |
| Applications |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification |
Polyclonal |
| Concentration |
1.0 mg/mL |
| Conjugate |
Unconjugated |
| Dilution |
Western Blot 1:250-1:1000, ELISA 1:1000-1:3000, Immunocytochemistry/Immunofluorescence 1:10-1:500 |
| Formulation |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide |
| Gene Accession No. |
Q7KZI7 |
| Gene Alias |
EC 2.7.11, EC 2.7.11.1, ELKL motif kinase, ELKL motif kinase 1, EMK-1, EMK1PAR-1, MAP/microtubule affinity-regulating kinase 2MGC99619, PAR1 homolog, Par1b, Ser/Thr protein kinase PAR-1B, serine/threonine protein kinase EMK, serine/threonine-protein kinase MARK2 |
| Gene Symbols |
MARK2 |
| Host Species |
Rabbit |
| Immunogen |
This affinity purified MARK2 [p Thr595] Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids surrounding T595 of Human MARK2 isoform a. The immunogen peptide was phosphorylated at the residue corresponding to T595. (Uniprot: Q7KZI7) |
| Molecular Weight of Antigen |
87.91 kDa |
| Purification Method |
Affinity Purified |
| Quantity |
0.1 mg |
| Regulatory Status |
RUO |
| Research Discipline |
Phospho Specific, Protein Kinase |
| Primary or Secondary |
Primary |
| Gene ID (Entrez) |
2011 |
| Test Specificity |
MARK2 pT595 affinity purified antibody is directed against the phosphorylated form of human MARK2 at the pT595 residue. The resultant affinity purified antibody was then cross-adsorbed against the non-phosphorylated form of the immunizing peptide. Reactivity occurs against human MARK2 pT595 protein and the antibody is specific for the phosphorylated form of the protein. Reactivity with non-phosphorylated human MARK2 is minimal by ELISA. Expect at least partial reactivity with other isoforms of MARK. The immunogen sequence is present in all forms identified to date and localizes to T595 on MARK2, T587 on MARK3, T591 on MARK1 and T568 on MARK4. A BLAST analysis was used to suggest reactivity with this protein from human, dog, bovine, mouse and rat sources based on 100% homology for the immunogen sequence. Cross reactivity with MARK2 homologues from other sources has not been determined. |
| Target Species |
Human, Mouse, Rat, Bovine, Canine |
| Content And Storage |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
| Isotype |
IgG |
|
Show More
Show Less
|
|