Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mark3 Antibody (OTI2G8), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. N272590A405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272590A405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272590A405 Supplier Novus Biologicals Supplier No. NBP272590AF405

Mouse Monoclonal Antibody

Mark3 Monoclonal specifically detects Mark3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Mark3
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2G8
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Cdc25C-associated protein kinase 1, cTAK1, C-TAK1, CTAK1ELKL motif kinase 2, EC 2.7.11, EC 2.7.11.1, EMK2, EMK-2, KP78, MAP/microtubule affinity-regulating kinase 3, PAR1A, Protein kinase STK10, Ser/Thr protein kinase PAR-1, Serine/threonine-protein kinase p78
Gene Symbols MARK3
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 473-729 of human Mark3 (NP_002367) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 4140.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.