Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MARK4 Antibody (OTI9B7), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. N272591A700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272591A700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272591A700 Supplier Novus Biologicals Supplier No. NBP272591AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MARK4 Monoclonal specifically detects MARK4 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.

Specifications

Antigen MARK4
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI9B7
Conjugate Alexa Fluor 700
Dilution Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 2.7.11, EC 2.7.11.1, FLJ12177, FLJ90097, KIAA1860MARK4L, MAP/microtubule affinity-regulating kinase 4, MAP/microtubule affinity-regulating kinase like 1, MAP/microtubule affinity-regulating kinase-like 1, MARK4 serine/threonine protein kinase, MARKL1L, MARKL1MARK4S, Nbla00650
Gene Symbols MARK4
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 390-467 of human MARK4 (NP_113605) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 57787.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.