Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MARS Polyclonal Antibody
Rabbit Polyclonal Antibody
$237.00 - $511.50
Specifications
Antigen | MARS |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119284
|
Invitrogen™
PA5119284 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121785
|
Invitrogen™
PA5121785 |
20 μL |
Each for $237.00
|
|
|||||
Description
MARS Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Methionyl-tRNA synthetase belongs to the class I family of tRNA synthetases. It catalyzes the chemical reaction:ATP + L-methionine + tRNA(Met) = AMP + diphosphate + L-methionyl-tRNA(Met).Specifications
MARS | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human, Mouse | |
MARS1 | |
CMT2U; cytosolic methionyl-tRNA synthetase; id:ibd5106; ILFS2; ILLD; MARS; mars protein; mars1; methionine tRNA ligase; methionine--tRNA ligase, cytoplasmic; methionine-tRNA synthetase; methionyl trna synthetase; methionyl-tRNA synthetase; methionyl-tRNA synthetase 1; methionyl-tRNA synthetase, cytoplasmic; metRS; MRS; Mtrns; SPG70; Unknown (protein for MGC:134096); zgc:66122 | |
MARS1 | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
P56192, Q68FL6 | |
216443, 4141 | |
Recombinant protein Methionyl tRNA Synthetase. The antigen corresponds to amino acid range 74-212 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title