Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MARVELD1 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB358652 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB358652 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB358652 Supplier Novus Biologicals Supplier No. NBP176243MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MARVELD1 Polyclonal antibody specifically detects MARVELD1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen MARVELD1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias bA548K23.8, FLJ22343, FLJ23440, GB14, MARVD1, MARVEL (membrane-associating) domain containing 1, MARVEL domain containing 1, MGC4415, MRVLDC1, putative MARVEL domain-containing protein 1, RP11-548K23.8
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide from near the amino terminus of human MARVELD1. The immunogen is located within the first 50 amino acids of MARVELD1. Amino Acid Squence: PPPRQPPPQARAARGA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 83742
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.