Learn More
Thermo Scientific™ MAS™ Diabetes Control and MAS Diabetes Max Control
Description
Monitor hemoglobin A1c methods with bi-level Thermo Scientific™ MAS™ Diabetes and MAS Diabetes Max Controls. These liquid, ready-to-use, assayed diabetes controls have been assayed for most major instrument systems. The MAS Diabetes MAX controls are packaged in convenient Load-and-Go tubes with barcodes1 provided for most major platforms, simplifying the daily QC routine and saving significant amounts of time.
- Human whole blood matrix
- Assayed values available for most major analyzers

Specifications
Specifications
Content And Storage | Two year shelf life at -25° to -15°C; 30 days open vial at 2° to 8°C |
Description | Vista Vials |
Detectable Analytes | Hemoglobin A1c |
Sample Type | Whole Blood |
Type | Control |
Level | Bi-level |
Packaging | 6 Pk. |
Quantity | 6 x 2 mL |
1Barcodes available for selected instruments.
Not all products are CE marked or have FDA clearance for sale in the U.S. Availability of products in each country depends on local regulatory marketing authorization status.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.