Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ MAS™ Omni·Infectious™ Quality Controls
Click to view available options
Description:
MAS HIV p24 Positive Control
MAS Infectious Negative Control
MAS Omni Infectious BSI Positive Control Panel
MAS Syphilis Positive Control
MAS anti-HBs Positive Control
Description
Assess the performance of Serological assays for viral and bacterial pathogens using the Thermo Scientific MAS Omni Infectious Quality Controls.
Assess the performance of serological assays for viral blood stream infections using the Thermo Scientific MAS Omni Infectious Quality Controls. These third-party control panels include individual positive controls for anti-HBs, HIV-p24 and Syphilis as well as consolidated HIV, HCV, HBV and HTLV for efficiency.
- 30 days open vial stability for Syphilis; anti-HBs and HIV p24 at 2 to 8°C
- Ready to use straight from the refrigerator
- Human serum matrix
- BSI Positive and Infectious Negative
- BSI Positive has Multiple analytes in a single vial
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | MAS Omni Infectious BSI Positive Control Panel |
| Detectable Analytes | HBsAg, anti-HIV, anti-HTLV, anti-HBc, anti-HCV, anti-HBs |
| Type | Control |
| Packaging | One box of control contains five tubes of positive control |
| Quantity | 5 x 5.0 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction