Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ MAS™ UrichemTRAK Controls
Click to view available options
Description:
UrichemTRAK Controls, Assayed
UrichemTRAK Controls, Assayed, Bi-level
Level:
1
2
Bi-level
Description
Monitor the most commonly tested urine chemistry analytes with liquid, assayed Thermo Scientific™ MAS™ UrichemTRAK Controls, available in a convenient Vista™ vial format. Ready-to-use and requiring no additional preparation, these controls will not interfere with ISEs.
- Liquid, human urine matrix
- Open and use; no equilibration required
- Two-year open-vial stability
- Individual levels or multi-pack configurations
- Stable for 24 months at 2° to 8°C
- Assayed values available for most major analyzers
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | UrichemTRAK Controls, Assayed, Bi-level |
| Detectable Analytes | Amylase, Calcium, Chloride, Cortisol, Creatinine, Glucose, hCG, Magnesium, Microalbumin, Osmolality, pH, Phosphorus, Potassium, Sodium, Specific Gravity, Total Protein, Urea Nitrogen, Uric Acid |
| Sample Type | Urine |
| Type | Control |
| Level | Bi-level |
| Packaging | 6 pack - 3 bottles of each level |
| Quantity | 15 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction