Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MASA Antibody, Novus Biologicals™
SDP

Catalog No. NBP1553620 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP1553620 20 μL
NBP155361 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP1553620 Supplier Novus Biologicals Supplier No. NBP15536120UL

Rabbit Polyclonal Antibody

MASA Polyclonal specifically detects MASA in Human samples. It is validated for Western Blot.

Specifications

Antigen MASA
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9UHY7
Gene Alias acireductone synthase, DKFZp586M0524, E1, EC 3.1.3.77, enolase-phosphatase 1, enolase-phosphatase E1, MASA homolog, MASAFLJ12594, MST1452,3-diketo-5-methylthio-1-phosphopentane phosphatase
Gene Symbols ENOPH1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to ENOPH1(enolase-phosphatase 1) Antibody(against the N terminal of ENOPH1. Peptide sequence IEENVKEYLQTHWEEEECQQDVSLLRKQAEEDAHLDGAVPIPAASGNGVD.
Molecular Weight of Antigen 29 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 58478
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.