Learn More
Mash1/Ascl1 Polyclonal Antibody, Rockland™

Goat Polyclonal Antibody
$174.73 - $636.24
Specifications
Antigen | Mash1/Ascl1 |
---|---|
Concentration | 1 mg/mL |
Applications | ELISA, Immunohistochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-1577
![]() ![]() |
Rockland Immunochemicals
600101MH4 |
100 μg |
Each for $636.24
|
|
|||||
50-199-1578
![]() ![]() |
Rockland Immunochemicals
600101MH4S |
25 μL |
Each for $174.73
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity purified antibody is directed against mouse Mash1/Ascl1. A BLAST analysis was used to suggest cross-reactivity with the antigen based on 100% homology with the immunizing sequence to mouse and rat, and 93% homology with human.
ASCL1 (also known as ASH1) is a basic helix-loop-helix transcription factor that is required for early development of the nervous system. Expressed in fetal brain, ASCL1 is essential for the proper development of autonomic neurons and for the survival of subsets of autonomic neurons. ASCL1 interaction with MEF-2A may regulate the expression of specific genes that are critical for the formation of distinct neuronal circuits within the central nervous system. The high level of ASCL1 expression in neuroendocrine tumors, such as medullary thyroid cancer, small cell lung cancer and lung cancer with neuroendocrine features may provide a useful marker for cancers with neuroendocrine features. Mapping to human chromosome 12, the ASCL1 gene contains a trinucleotide repeat region, making this locus a candidate for inherited disease.Specifications
Mash1/Ascl1 | |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
Goat | |
Human, Mouse | |
P50553, Q02067 | |
17172, 429 | |
Primary | |
Antibody |
1 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
achaete scute protein; achaete-scute complex homolog 1; achaete-scute complex homolog 1 (Drosophila); achaete-scute complex homolog-like 1; achaete-scute complex-like 1; achaete-scute complex-like protein 1; achaete-scute family bHLH transcription factor 1; Achaete-scute homolog 1; achaete-scute homolog 1-like; AI225900; ASCL1; ASH1; ASH-1; BHLHA46; Class A basic helix-loop-helix protein 46; HASH1; KMT2H; LOC100342114; mammalian achaete scute homolog 1; Mash1; Mash-1 | |
Ascl1 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Ascl1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.