Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mast Cell Marker Antibody (MCG35), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP250484R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-50484R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-50484R Supplier Novus Biologicals Supplier No. NBP250484R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mast Cell Marker Monoclonal specifically detects Mast Cell Marker in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen Mast Cell Marker
Applications Immunohistochemistry
Classification Monoclonal
Clone MCG35
Conjugate DyLight 550
Dilution Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Mouse
Immunogen Spleen cells and bone marrow cells (erythrocyte depleted) from a patient with systemic mastocytosis. The bone marrow preparation consisted of 50% typical mast cells.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Test Specificity The antibody does not react with basophils or other haemopoietic cell types.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.