Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAST205 Antibody (OTI3A5), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP27292A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27292A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27292A94 Supplier Novus Biologicals Supplier No. NBP272592AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MAST205 Monoclonal specifically detects MAST205 in Human, Mouse, Rat samples. It is validated for Western Blot, Knockout Validated.

Specifications

Antigen MAST205
Applications Western Blot, Immunoassay
Classification Monoclonal
Clone OTI3A5
Conjugate Alexa Fluor 594
Dilution Western Blot, Knockout Validated
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias EC 2.7.11, EC 2.7.11.1, FLJ39200, KIAA0807RP4-533D7.1, MAST205microtubule associated testis specific serine/threonine protein kinase, microtubule associated serine/threonine kinase 2, microtubule-associated serine/threonine-protein kinase 2, MTSSK
Gene Symbols MAST2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1482-1798 of human MAST205 (NP_055927) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 23139
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.