Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MASTL Antibody, Novus Biologicals™
SDP

Catalog No. NBP122987 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP122987 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP122987 Supplier Novus Biologicals Supplier No. NBP122987
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MASTL Polyclonal specifically detects MASTL in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen MASTL
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry 1:1000 - 1:5000, Immunoprecipitation 2-10 ug/mg lysate, Immunohistochemistry-Paraffin 1:1000 - 1:5000
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. Q96GX5
Gene Alias FLJ14813EC 2.7.11.1, GREATWALL, GW, GWL, hGWL, MAST-L, microtubule associated serine/threonine kinase-like, Microtubule-associated serine/threonine-protein kinase-like, RP11-85G18.2, THC2greatwall protein kinase
Gene Symbols MASTL
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 550 and 600 of human microtubule associated serine/threonine kinase-like using the numbering given in entry NP_116233.2 (GeneID 84930).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 84930
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.