Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MASTL Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$167.95 - $630.77
Specifications
| Antigen | MASTL |
|---|---|
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2450
|
Bethyl Laboratories, Inc
IHC00638T |
10 μL |
Each for $167.95
|
|
|||||
50-157-2449
|
Bethyl Laboratories, Inc
IHC00638 |
100 μL |
Each for $630.77
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
Serine/threonine kinase that plays a key role in M phase by acting as a regulator of mitosis entry and maintece. Acts by promoting the inactivation of protein phosphatase 2A (PP2A) during M phase: does not directly inhibit PP2A but acts by mediating phosphorylation and subsequent activation of ARPP19 and ENSA at 'Ser-62' and 'Ser-67', respectively. ARPP19 and ENSA are phosphatase inhibitors that specifically inhibit the PPP2R2D (PR55-delta) subunit of PP2A. Inactivation of PP2A during M phase is essential to keep cyclin-B1-CDK1 activity high. Following DNA damage, it is also involved in checkpoint recovery by being inhibited. Phosphorylates histone protein in vitro; however such activity is unsure in vivo. May be involved in megakaryocyte differentiation. Ota T., Nat. Genet. 36:40-45(2004). Deloukas P., Nature 429:375-381(2004). Beausoleil S.A., Nat. Biotechnol. 24:1285-1292(2006).Specifications
| MASTL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| GREATWALL, GW, GWL, MAST-L, RP11-85G18.2, THC2, hGWL, greatwall protein kinase, microtubule-associated serine/threonine-protein kinase-like, serine/threonine-protein kinase greatwall | |
| MASTL | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
| Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q96GX5 | |
| 84930 | |
| Between 550 and 600 | |
| Primary | |
| 4° C | |
| MASTL |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title