Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAT1/2A Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. N1094162A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1094162A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1094162A30 Supplier Novus Biologicals Supplier No. NB11094162AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MAT1/2A Polyclonal specifically detects MAT1/2A in Human, Rat, Bovine, Primate, Zebrafish, Mouse (Negative) samples. It is validated for Western Blot.

Specifications

Antigen MAT1/2A
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias AdoMet synthase 2, AMS2, EC 2.5.1.6, MAT 2, MATA2SAMS2adoMet synthetase 2, MATII, MAT-II, Methionine adenosyltransferase 2, Methionine adenosyltransferase II, methionine adenosyltransferase II, alpha, S-adenosylmethionine synthase isoform type-2, S-adenosylmethionine synthetase isoform type-2
Gene Symbols MAT2A
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human MAT2A protein (within residues 100-200). [Swiss-ProtNo. P31153]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 4144
Target Species Human, Rat, Bovine, Primate, Zebrafish, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.