Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAT1A Antibody, DyLight 680, Novus Biologicals™
SDP

Catalog No. NB044072 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB044072 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB044072 Supplier Novus Biologicals Supplier No. NBP297484FR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MAT1A Polyclonal specifically detects MAT1A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MAT1A
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 680
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias AdoMet synthase 1, adoMet synthetase 1, AMS1, MAT, MATA1MAT 1, MAT-I/III, Methionine adenosyltransferase 1, methionine adenosyltransferase I, alpha, Methionine adenosyltransferase I/III, S-adenosylmethionine synthase isoform type-1, S-adenosylmethionine synthetase isoform type-1, SAMS, SAMS1EC 2.5.1.6
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human MAT1A fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 4143
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.