Learn More
MAT1A Rabbit anti-Human, Mouse, Plant, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | MAT1A |
---|---|
Concentration | 0.15 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-7954
![]() ![]() |
Proteintech Group Inc
123951AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody was raised against the full length of human MAT1A and may cross-react with MAT2A. Western blot analysis using this antibody detected a single band around 44-50kD in liver.
MAT1A catalyzes the formation of S-adenosylmethionine from methionine and ATP. Methionine adenosyltransferase deficiency is caused by recessive and domit mutations, the latter identified in autosomal domit persistant hypermethioninemia.This gne encodes methionine adenosyltransferase I (alpha isoform), which catalyzes the formation of S-adenosylmethionine from methionine and ATP. Methionine adenosyltransferase deficiency is caused by recessive and domit mutations, the latter identified in autosomal domit persistant hypermethioninemia.Specifications
MAT1A | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Plant, Rat | |
P13444, Q00266, Q91X83 | |
11720, 25331, 4143 | |
MAT1A Fusion Protein Ag3076 | |
Primary | |
-20°C | |
MAT1A |
0.15 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
AdoMet synthase 1, AMS1, MAT, MAT 1, MAT I/III, MAT1A, MATA1, SAMS, SAMS1 | |
MAT1A | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.