Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAT2A Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $487.50
Specifications
Antigen | MAT2A |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1.0 ug/ml, Simple Western, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:2000, Immunoprecipitation reported in scientific literature (PMID 25294683), Immunohistochemistry-Paraffin 1:200 |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11094158T
![]() |
Novus Biologicals
NB11094158SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB11094158
![]() |
Novus Biologicals
NB11094158 |
0.1 mL |
Each for $487.50
|
|
|||||
Description
MAT2A Polyclonal specifically detects MAT2A in Human, Mouse, Rat, Porcine, Bovine, Primate samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
MAT2A | |
Western Blot 1.0 ug/ml, Simple Western, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:2000, Immunoprecipitation reported in scientific literature (PMID 25294683), Immunohistochemistry-Paraffin 1:200 | |
Polyclonal | |
Rabbit | |
Amino Acids Drugs and other small molecules, Cancer, Lipid and Metabolism, Signal Transduction | |
PBS with 0.02% Sodium Azide | |
AdoMet synthase 2, AMS2, EC 2.5.1.6, MAT 2, MATA2SAMS2adoMet synthetase 2, MATII, MAT-II, Methionine adenosyltransferase 2, Methionine adenosyltransferase II, methionine adenosyltransferase II, alpha, S-adenosylmethionine synthase isoform type-2, S-adenosylmethionine synthetase isoform type-2 | |
MAT2A | |
IgG | |
Affinity Purified | |
Human, rat, mouse, orangutan, bovine and monkey. Immunogen sequence has 94% homology to porcine and chicken and 86% homology to Zebrafish. |
1.0 mg/mL | |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
RUO | |
Human, Mouse, Rat, Pig, Bovine, Primate | |
P31153 | |
4144 | |
Synthetic peptide made to an N-terminal portion of human MAT2A (within residues 1-100). [Swiss-Prot# P31153] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
43 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title