Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAT2A Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB338967 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB338967 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB338967 Supplier Novus Biologicals Supplier No. NB11094158PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MAT2A Polyclonal antibody specifically detects MAT2A in Human, Mouse, Rat, Porcine, Bovine, Primate samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen MAT2A
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias AdoMet synthase 2, AMS2, EC 2.5.1.6, MAT 2, MATA2SAMS2adoMet synthetase 2, MATII, MAT-II, Methionine adenosyltransferase 2, Methionine adenosyltransferase II, methionine adenosyltransferase II, alpha, S-adenosylmethionine synthase isoform type-2, S-adenosylmethionine synthetase isoform type-2
Host Species Rabbit
Immunogen Synthetic peptide made to an N-terminal portion of human MAT2A (within residues 1-100). [Swiss-Prot# P31153]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Cancer, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4144
Target Species Human, Mouse, Rat, Pig, Bovine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.