Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MATH1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP145693 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP145693 0.1 mg
NB018576 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP145693 Supplier Novus Biologicals Supplier No. NBP145693

Rabbit Polyclonal Antibody

MATH1 Polyclonal specifically detects MATH1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen MATH1
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry
Formulation PBS (pH 7.2), 0.5% BSA, 5 mM EDTA, 30% Glycerol with 0.03% Proclin 300
Gene Accession No. Q92858
Gene Alias ATH1protein atonal homolog 1, atonal homolog 1 (Drosophila), BHLHA14, bHLHa14Math1, Class A basic helix-loop-helix protein 14, hATH1, Helix-loop-helix protein hATH-1, MATH-1
Gene Symbols ATOH1
Host Species Rabbit
Immunogen Synthetic peptide surrounding amino acid 190 of human Math1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 474
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.